siRNA Details
Gene Symbol: LOC101928449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110089.1 | 1 | 289 | |||
GenBank mRNA | BC031963.1 | - | 1 | 289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110089.1 | 1 | 289 | |||
GenBank mRNA | BC031963.1 | - | 1 | 289 |