siRNA Details
| Gene Symbol: CACTIN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038865.1 | 4 | 1887 | |||
| GenBank mRNA | AK097999.1 | - | 4 | 1888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CACTIN-AS1| Gene Symbol: CACTIN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038865.1 | 4 | 1887 | |||
| GenBank mRNA | AK097999.1 | - | 4 | 1888 | ||