siRNA Details
Gene Symbol: CACTIN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038865.1 | 3 | 508 | |||
GenBank mRNA | AK097999.1 | - | 3 | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB116596.1 | - | 3 | 509 | Chr. 19 - 3607247 - 3612077 [+] on Build GRCh38 |
