siRNA Details
| Gene Symbol: LOC102723322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134311.1 | 3 | 403 | |||
| GenBank mRNA | BX094626.1 | - | 3 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723322| Gene Symbol: LOC102723322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134311.1 | 3 | 403 | |||
| GenBank mRNA | BX094626.1 | - | 3 | 403 | ||