siRNA Details
| Gene Symbol: LOC283887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135196.1 | 2 | 1739 | |||
| NR_135197.1 | 2 | 1725 | ||||
| GenBank mRNA | AK093382.1 | - | 2 | 1588 | ||
| BC023651.2 | - | 2 | 1675 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283887| Gene Symbol: LOC283887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135196.1 | 2 | 1739 | |||
| NR_135197.1 | 2 | 1725 | ||||
| GenBank mRNA | AK093382.1 | - | 2 | 1588 | ||
| BC023651.2 | - | 2 | 1675 | |||