siRNA Details
| Gene Symbol: LINC01589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131244.1 | 1 | 158 | |||
| GenBank mRNA | BC045163.1 | - | 1 | 132 | ||
| BQ188486.1 | - | 1 | 158 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG122316.1 | - | 1 | 131 | Chr. 22 - 45604448 - 45605620 [-] on Build GRCh38 | |

LINC01589
Genomic Map