siRNA Details
Gene Symbol: LOC105377267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136191.1 | 6 | 2911 | |||
GenBank mRNA | AK124272.1 | - | 4 | 2586 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105377267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136191.1 | 6 | 2911 | |||
GenBank mRNA | AK124272.1 | - | 4 | 2586 |