siRNA Details
| Gene Symbol: LOC340512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126029.1 | 2 | 273 | |||
| GenBank mRNA | AK097706.1 | - | 2 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC340512| Gene Symbol: LOC340512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126029.1 | 2 | 273 | |||
| GenBank mRNA | AK097706.1 | - | 2 | 273 | ||