siRNA Details
| Gene Symbol: TMEM72-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033842.1 | 8 | 3202 | |||
| GenBank mRNA | AK056518.1 | - | 8 | 3203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM72-AS1| Gene Symbol: TMEM72-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033842.1 | 8 | 3202 | |||
| GenBank mRNA | AK056518.1 | - | 8 | 3203 | ||