siRNA Details
| Gene Symbol: LINC00906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027318.2 | 2 | 3059 | |||
| GenBank mRNA | BX647231.1 | - | 2 | 3057 | ||
| DB513590.2 | - | 1 | 314 | |||
| Gene Symbol: LOC102724958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110760.1 | 1 | 296 | |||
| GenBank mRNA | AY726575.1 | - | 1 | 296 | ||

LINC00906
Genomic Map