siRNA Details
| Gene Symbol: LINC00518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027793.1 | 3 | 952 | |||
| GenBank mRNA | BC016666.1 | - | 3 | 952 | ||
| BC017496.1 | - | 3 | 671 | 3921932 | ||
| BC028118.1 | - | 3 | 682 | 3922009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00518| Gene Symbol: LINC00518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027793.1 | 3 | 952 | |||
| GenBank mRNA | BC016666.1 | - | 3 | 952 | ||
| BC017496.1 | - | 3 | 671 | 3921932 | ||
| BC028118.1 | - | 3 | 682 | 3922009 | ||