siRNA Details
Gene Symbol: HOXA-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038831.1 | 3 | 1805 | |||
NR_038832.1 | 3 | 1731 | ||||
GenBank mRNA | AK091933.1 | - | 3 | 1810 | ||
AK096334.1 | - | 1 | 1037 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HOXA-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038831.1 | 3 | 1805 | |||
NR_038832.1 | 3 | 1731 | ||||
GenBank mRNA | AK091933.1 | - | 3 | 1810 | ||
AK096334.1 | - | 1 | 1037 |