siRNA Details
| Gene Symbol: LOC653160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037869.1 | 3 | 1782 | |||
| GenBank mRNA | AF085941.1 | - | 1 | 387 | ||
| BC021230.2 | - | 3 | 1782 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 LOC653160
LOC653160| Gene Symbol: LOC653160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037869.1 | 3 | 1782 | |||
| GenBank mRNA | AF085941.1 | - | 1 | 387 | ||
| BC021230.2 | - | 3 | 1782 | |||
 Back To Top
 Back To Top