siRNA Details
Gene Symbol: LINC01326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG190199.1 | - | 2 | 279 | ||
BG199333.1 | - | 2 | 289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG190199.1 | - | 2 | 279 | ||
BG199333.1 | - | 2 | 289 |