siRNA Details
| Gene Symbol: LVCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131958.1 | 2 | 518 | |||
| NR_131959.1 | 2 | 411 | ||||
| GenBank mRNA | AL038572.2 | - | 2 | 410 | ||
| BU569738.1 | - | 2 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LVCAT1| Gene Symbol: LVCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131958.1 | 2 | 518 | |||
| NR_131959.1 | 2 | 411 | ||||
| GenBank mRNA | AL038572.2 | - | 2 | 410 | ||
| BU569738.1 | - | 2 | 330 | |||