siRNA Details
| Gene Symbol: ZNF648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000302.1 | XP_016855791.1 | 1 | 664 | ||
| Gene Symbol: LINC01344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128015.1 | - | 4 | 337 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB059015.1 | - | 4 | 337 | Chr. 1 - 182203929 - 182244399 [-] on Build GRCh38 | |

ZNF648
Genomic Map