siRNA Details
| Gene Symbol: DIP2A-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046400.1 | 4 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIP2A-IT1| Gene Symbol: DIP2A-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046400.1 | 4 | 352 | |||