siRNA Details
| Gene Symbol: UBL7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038448.1 | 5 | 1656 | |||
| NR_038449.1 | 4 | 1295 | ||||
| NR_120377.1 | 4 | 1291 | ||||
| GenBank mRNA | BX104607.1 | - | 4 | 660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBL7-AS1| Gene Symbol: UBL7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038448.1 | 5 | 1656 | |||
| NR_038449.1 | 4 | 1295 | ||||
| NR_120377.1 | 4 | 1291 | ||||
| GenBank mRNA | BX104607.1 | - | 4 | 660 | ||