siRNA Details
| Gene Symbol: LOC101928100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120430.1 | 4 | 2169 | |||
| GenBank mRNA | AK096314.1 | - | 4 | 2169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928100| Gene Symbol: LOC101928100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120430.1 | 4 | 2169 | |||
| GenBank mRNA | AK096314.1 | - | 4 | 2169 | ||