siRNA Details
| Gene Symbol: LOC100996419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134283.1 | 2 | 981 | |||
| GenBank mRNA | BC040901.1 | - | 2 | 971 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996419| Gene Symbol: LOC100996419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134283.1 | 2 | 981 | |||
| GenBank mRNA | BC040901.1 | - | 2 | 971 | ||