siRNA Details
| Gene Symbol: LOC100996419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134283.1 | 2 | 421 | |||
| GenBank mRNA | BC040901.1 | - | 2 | 411 | ||
| CN357255.1 | - | 2 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996419| Gene Symbol: LOC100996419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134283.1 | 2 | 421 | |||
| GenBank mRNA | BC040901.1 | - | 2 | 411 | ||
| CN357255.1 | - | 2 | 435 | |||