siRNA Details
Gene Symbol: LINC00661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026828.1 | 7 | 721 | |||
GenBank mRNA | AK128240.1 | - | 2 | 219 | ||
BC117476.1 | - | 3 | 323 | |||
BC144048.1 | - | 3 | 323 | |||
CR749400.1 | - | 7 | 721 |
