siRNA Details
| Gene Symbol: MGC45922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038359.1 | 3 | 376 | |||
| GenBank mRNA | BC033237.1 | - | 3 | 376 | 5428578 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC45922| Gene Symbol: MGC45922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038359.1 | 3 | 376 | |||
| GenBank mRNA | BC033237.1 | - | 3 | 376 | 5428578 | |