siRNA Details
| Gene Symbol: LOC102723385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119379.1 | 3 | 377 | |||
| NR_119380.1 | 2 | 309 | ||||
| NR_119381.1 | 2 | 279 | ||||
| NR_119382.1 | 2 | 334 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723385| Gene Symbol: LOC102723385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119379.1 | 3 | 377 | |||
| NR_119380.1 | 2 | 309 | ||||
| NR_119381.1 | 2 | 279 | ||||
| NR_119382.1 | 2 | 334 | ||||