siRNA Details
Gene Symbol: LOC107984745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022786.1 | XP_016878275.1 | 2 | 731 |
Gene Symbol: LOC101929767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093551.1 | - | 1 | 1006 |
