siRNA Details
| Gene Symbol: LINC00927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033833.1 | 3 | 2274 | |||
| GenBank mRNA | BC040586.1 | - | 3 | 2237 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00927| Gene Symbol: LINC00927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033833.1 | 3 | 2274 | |||
| GenBank mRNA | BC040586.1 | - | 3 | 2237 | ||