siRNA Details
| Gene Symbol: CTD-3080P12.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109911.1 | 2 | 392 | |||
| GenBank mRNA | AK091791.1 | - | 2 | 392 | ||
| BC032469.1 | - | 2 | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTD-3080P12.3| Gene Symbol: CTD-3080P12.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109911.1 | 2 | 392 | |||
| GenBank mRNA | AK091791.1 | - | 2 | 392 | ||
| BC032469.1 | - | 2 | 379 | |||