siRNA Details
| Gene Symbol: LOC100132735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038399.1 | 3 | 434 | |||
| GenBank mRNA | BC039503.1 | - | 3 | 463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100132735| Gene Symbol: LOC100132735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038399.1 | 3 | 434 | |||
| GenBank mRNA | BC039503.1 | - | 3 | 463 | ||