siRNA Details
| Gene Symbol: LOC105372897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135085.1 | 2 | 137 | |||
| GenBank mRNA | BX119711.1 | - | 2 | 137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372897| Gene Symbol: LOC105372897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135085.1 | 2 | 137 | |||
| GenBank mRNA | BX119711.1 | - | 2 | 137 | ||