siRNA Details
| Gene Symbol: LINC01168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046231.1 | 3 | 4587 | |||
| GenBank mRNA | AK125849.1 | - | 3 | 4587 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01168| Gene Symbol: LINC01168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046231.1 | 3 | 4587 | |||
| GenBank mRNA | AK125849.1 | - | 3 | 4587 | ||