siRNA Details
| Gene Symbol: DRAIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026979.1 | 1 | 69 | |||
| GenBank mRNA | BC029061.1 | - | 1 | 69 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI762341.1 | - | 1 | 68 | Chr. 15 - 69561723 - 69565363 [+] on Build GRCh38 | |

DRAIC
Genomic Map