siRNA Details
| Gene Symbol: LINC00347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034024.1 | 3 | 694 | |||
| NR_034025.1 | 3 | 650 | ||||
| GenBank mRNA | BC043259.2 | - | 3 | 687 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU854065.1 | - | 3 | 653 | Chr. 13 - 74552843 - 74555731 [+] on Build GRCh38 | |

LINC00347
Genomic Map