siRNA Details
| Gene Symbol: LINC00347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034024.1 | 3 | 1368 | |||
| NR_034025.1 | 3 | 1324 | ||||
| GenBank mRNA | BC043259.2 | - | 3 | 1361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00347| Gene Symbol: LINC00347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034024.1 | 3 | 1368 | |||
| NR_034025.1 | 3 | 1324 | ||||
| GenBank mRNA | BC043259.2 | - | 3 | 1361 | ||