siRNA Details
| Gene Symbol: LOC105369739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135028.1 | 4 | 578 | |||
| GenBank mRNA | BX096171.1 | - | 4 | 578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369739| Gene Symbol: LOC105369739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135028.1 | 4 | 578 | |||
| GenBank mRNA | BX096171.1 | - | 4 | 578 | ||