siRNA Details
| Gene Symbol: LINC00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128266.1 | - | 4 | 1068 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00514| Gene Symbol: LINC00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128266.1 | - | 4 | 1068 | ||