siRNA Details
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 4 | 1029 | |||
GenBank mRNA | AK094518.1 | - | 4 | 491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA133886.1 | - | 4 | 492 | Chr. 5 - 60488251 - 60546255 [+] on Build GRCh38 |
