siRNA Details
| Gene Symbol: LOC105379194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134244.1 | 2 | 610 | |||
| NR_134245.1 | 2 | 466 | ||||
| GenBank mRNA | AI792600.1 | - | 2 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105379194| Gene Symbol: LOC105379194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134244.1 | 2 | 610 | |||
| NR_134245.1 | 2 | 466 | ||||
| GenBank mRNA | AI792600.1 | - | 2 | 466 | ||