siRNA Details
| Gene Symbol: LOC102724434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130921.1 | 4 | 1966 | |||
| NR_130922.1 | 4 | 1948 | ||||
| NR_130923.1 | 3 | 1927 | ||||
| GenBank mRNA | AK097428.1 | - | 3 | 1927 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724434| Gene Symbol: LOC102724434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130921.1 | 4 | 1966 | |||
| NR_130922.1 | 4 | 1948 | ||||
| NR_130923.1 | 3 | 1927 | ||||
| GenBank mRNA | AK097428.1 | - | 3 | 1927 | ||