siRNA Details
| Gene Symbol: ZNF528-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125345.1 | 1 | 111 | |||
| GenBank mRNA | AK097590.1 | - | 1 | 119 | ||
| BI755704.1 | - | 1 | 120 | |||
| BX537909.1 | - | 1 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF528-AS1| Gene Symbol: ZNF528-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125345.1 | 1 | 111 | |||
| GenBank mRNA | AK097590.1 | - | 1 | 119 | ||
| BI755704.1 | - | 1 | 120 | |||
| BX537909.1 | - | 1 | 86 | |||