siRNA Details
| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126016.1 | 3 | 954 | |||
| NR_126017.1 | 3 | 872 | ||||
| NR_126018.1 | 2 | 859 | ||||
| NR_126019.1 | 2 | 941 | ||||
| GenBank mRNA | BC042032.1 | - | 2 | 859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYRM4-AS1| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126016.1 | 3 | 954 | |||
| NR_126017.1 | 3 | 872 | ||||
| NR_126018.1 | 2 | 859 | ||||
| NR_126019.1 | 2 | 941 | ||||
| GenBank mRNA | BC042032.1 | - | 2 | 859 | ||