siRNA Details
| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126010.1 | 2 | 232 | |||
| NR_126011.1 | 3 | 281 | ||||
| GenBank mRNA | AK093119.1 | - | 2 | 232 | ||
| BC039446.1 | - | 2 | 199 | |||
| CD515480.1 | - | 3 | 281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284825| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126010.1 | 2 | 232 | |||
| NR_126011.1 | 3 | 281 | ||||
| GenBank mRNA | AK093119.1 | - | 2 | 232 | ||
| BC039446.1 | - | 2 | 199 | |||
| CD515480.1 | - | 3 | 281 | |||