siRNA Details
Gene Symbol: LINC01554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026936.1 | 3 | 829 | |||
GenBank mRNA | AK096140.1 | - | 4 | 728 | ||
AY168789.1 | - | 4 | 708 | |||
BC093643.1 | - | 2 | 338 | |||
BC101577.1 | - | 2 | 338 |
