siRNA Details
| Gene Symbol: LOC101929054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135307.1 | 2 | 982 | |||
| GenBank mRNA | BC039681.1 | - | 2 | 956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929054| Gene Symbol: LOC101929054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135307.1 | 2 | 982 | |||
| GenBank mRNA | BC039681.1 | - | 2 | 956 | ||