siRNA Details
| Gene Symbol: LOC100506107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134683.1 | 2 | 392 | |||
| GenBank mRNA | AF086144.1 | - | 2 | 392 | ||
| BU078148.1 | - | 2 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506107| Gene Symbol: LOC100506107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134683.1 | 2 | 392 | |||
| GenBank mRNA | AF086144.1 | - | 2 | 392 | ||
| BU078148.1 | - | 2 | 385 | |||