siRNA Details
| Gene Symbol: ZNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL583605.3 | - | 2 | 663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF165| Gene Symbol: ZNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL583605.3 | - | 2 | 663 | ||