siRNA Details
| Gene Symbol: C5orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277348.1 | NP_001264277.1 | 6 | 1070 | ||
| GenBank mRNA | AK026965.1 | - | 6 | 1070 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf66| Gene Symbol: C5orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277348.1 | NP_001264277.1 | 6 | 1070 | ||
| GenBank mRNA | AK026965.1 | - | 6 | 1070 | ||