siRNA Details
Gene Symbol: DIAPH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046539.2 | 4 | 470 | |||
NR_051993.2 | 3 | 310 | ||||
NR_051994.2 | 4 | 398 | ||||
NR_109838.1 | 5 | 589 | ||||
GenBank mRNA | BX093461.1 | - | 4 | 469 | ||
BX398653.2 | - | 5 | 583 |
