siRNA Details
| Gene Symbol: LINC01477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110791.1 | 1 | 159 | |||
| GenBank mRNA | BC034625.1 | - | 1 | 159 | ||
| BC045816.1 | - | 1 | 159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01477| Gene Symbol: LINC01477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110791.1 | 1 | 159 | |||
| GenBank mRNA | BC034625.1 | - | 1 | 159 | ||
| BC045816.1 | - | 1 | 159 | |||