siRNA Details
Gene Symbol: LINC01477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110791.1 | 3 | 933 | |||
GenBank mRNA | BC034625.1 | - | 3 | 933 | ||
BC045816.1 | - | 3 | 933 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110791.1 | 3 | 933 | |||
GenBank mRNA | BC034625.1 | - | 3 | 933 | ||
BC045816.1 | - | 3 | 933 |