siRNA Details
| Gene Symbol: LOC100131315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109767.1 | 2 | 382 | |||
| GenBank mRNA | AK093166.1 | - | 1 | 704 | ||
| AW582922.1 | - | 2 | 382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100131315| Gene Symbol: LOC100131315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109767.1 | 2 | 382 | |||
| GenBank mRNA | AK093166.1 | - | 1 | 704 | ||
| AW582922.1 | - | 2 | 382 | |||